Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Zymolyase - 20T from Arthrobacter luteus

Catalog No. MP08320921 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08320921 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP08320921 Supplier MP Biomedicals™ Supplier No. 08320921
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zymolyase-20T is an enzyme preparation from a submerged culture of Arthrobacter luteus which effectively lyses cell walls of viable yeast cells.

  • Preparation is a lyophilized powder prepared by salting out from the culture fluid with ammonium sulfate and contains 20,000 units enzyme/g
  • Primary enzyme in this preparation responsible for lysis of viable yeast cells is ß-1,3-glucan laminaripentaohydrolase
  • Hydrolyzes glucose polymers at the ß-1,3-glucan linkages releasing laminaripentaose as the principal product
TRUSTED_SUSTAINABILITY

Specifications

Concentration 20,000 U/g
For Use With (Application) Yeast Cell Culture, Cell Structure
Content And Storage 2°C to 8°C
Source Arthrobacter luteus
Quantity 1 g
Product Type Zymolase
Form Lyophilized Powder

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.