Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Zyxin Monoclonal Antibody (CL2502)
GREENER_CHOICE

Catalog No. PIMA531367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531367 Supplier Invitrogen™ Supplier No. MA531367
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: PAPKFSPVTP KFTPVASKFS PGAPGGSGSQ PNQKLGHPEA LSAGTGSPQP PSFTYAQQRE KPRVQEKQHP VPPPAQNQNQ VRSPGAPGPL TLKEVEELEQ LTQQLMQDME HPQRQNVAVN E Highest antigen sequence indentity to the following orthologs: Rat - 85%, Mouse - 84%.

Zyxin is involved in focal adhesions, which are actin-rich structures that enable cells to adhere to the extracellular matrix and at which protein complexes involved in signal transduction assemble. Zyxin is a zinc-binding phosphoprotein that concentrates at focal adhesions and along the actin cytoskeleton. Zyxin has an N-terminal proline-rich domain and three LIM domains in its C-terminal half. The proline-rich domain may interact with SH3 domains of proteins involved in signal transduction pathways while the LIM domains are likely involved in protein-protein binding. Zyxin may function as a messenger in the signal transduction pathway that mediates adhesion-stimulated changes in gene expression and may modulate the cytoskeletal organization of actin bundles. Alternative splicing results in multiple transcript variants that encode the same isoform.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zyxin
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone CL2502
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZYX
Gene Accession No. Q15942
Gene Alias 9530098H06Rik; ESP-2; HED-2; LOW QUALITY PROTEIN: zyxin; R75157; ZYX; Zyxin; zyxin-2
Gene Symbols ZYX
Host Species Mouse
Immunogen Recombinant protein fragment of Human ZYX (Product #RP-102436)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7791
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.