Assay Details
| Gene Symbol: MED14_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_315707.4 | XP_315707.4 | 2 - 3 | 1591 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED14_ANOGA| Gene Symbol: MED14_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_315707.4 | XP_315707.4 | 2 - 3 | 1591 | 61 | |
Set Membership: |
|