Assay Details
| Gene Symbol: MED14_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_315707.4 | XP_315707.4 | 3 - 4 | 3707 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED14_ANOGA| Gene Symbol: MED14_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_315707.4 | XP_315707.4 | 3 - 4 | 3707 | 64 | |
Set Membership: |
|