Assay Details
| Gene Symbol: HEXA_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_315780.4 | XP_315780.4 | 2 - 3 | 219 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HEXA_ANOGA| Gene Symbol: HEXA_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_315780.4 | XP_315780.4 | 2 - 3 | 219 | 60 | |
Set Membership: |
|