Assay Details
| Gene Symbol: MED24_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_315936.4 | XP_315936.4 | 2 - 3 | 261 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED24_ANOGA| Gene Symbol: MED24_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_315936.4 | XP_315936.4 | 2 - 3 | 261 | 61 | |
Set Membership: |
|