Assay Details
| Gene Symbol: KMO_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_315983.4 | XP_315983.2 | 1 - 2 | 317 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KMO_ANOGA| Gene Symbol: KMO_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_315983.4 | XP_315983.2 | 1 - 2 | 317 | 81 | |
Set Membership: |
|