Assay Details
| Gene Symbol: OBP49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_316128.3 | XP_316128.3 | 1 - 2 | 101 | 51 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OBP49| Gene Symbol: OBP49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_316128.3 | XP_316128.3 | 1 - 2 | 101 | 51 | |
Set Membership: |
|