Assay Details
| Gene Symbol: OBP49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_316128.3 | XP_316128.3 | 2 - 3 | 217 | 60 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

OBP49| Gene Symbol: OBP49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_316128.3 | XP_316128.3 | 2 - 3 | 217 | 60 | |
Set Membership: | 
		   
					
						
							 |