Assay Details
| Gene Symbol: OBP56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_317953.2 | XP_317953.2 | 2 - 3 | 331 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OBP56| Gene Symbol: OBP56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_317953.2 | XP_317953.2 | 2 - 3 | 331 | 65 | |
Set Membership: |
|