Assay Details
| Gene Symbol: 1278597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_318214.4 | XP_318214.4 | 1 - 2 | 199 | 136 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

1278597| Gene Symbol: 1278597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_318214.4 | XP_318214.4 | 1 - 2 | 199 | 136 | |
Set Membership: |
|