Assay Details
| Gene Symbol: 1278620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_318239.4 | XP_318239.4 | 16 - 17 | 2501 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

1278620| Gene Symbol: 1278620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_318239.4 | XP_318239.4 | 16 - 17 | 2501 | 79 | |
Set Membership: |
|