Assay Details
| Gene Symbol: MED6_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_319180.4 | XP_319180.3 | 2 - 3 | 556 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED6_ANOGA| Gene Symbol: MED6_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_319180.4 | XP_319180.3 | 2 - 3 | 556 | 77 | |
Set Membership: |
|