Assay Details
| Gene Symbol: 1279737 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_319502.3 | XP_319502.1 | 5 - 6 | 1087 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

1279737| Gene Symbol: 1279737 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_319502.3 | XP_319502.1 | 5 - 6 | 1087 | 76 | |
Set Membership: |
|