Assay Details
| Gene Symbol: MED15_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_553774.3 | XP_553774.3 | 3 - 4 | 626 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED15_ANOGA| Gene Symbol: MED15_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_553774.3 | XP_553774.3 | 3 - 4 | 626 | 74 | |
Set Membership: |
|