Assay Details
| Gene Symbol: 3292445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_554502.2 | XP_554502.2 | 3 - 4 | 742 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

3292445| Gene Symbol: 3292445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_554502.2 | XP_554502.2 | 3 - 4 | 742 | 67 | |
Set Membership: |
|