Assay Details
| Gene Symbol: RL5_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_319782.5 | XP_319782.3 | 1 - 2 | 700 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RL5_ANOGA| Gene Symbol: RL5_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_319782.5 | XP_319782.3 | 1 - 2 | 700 | 74 | |
Set Membership: |
|