Assay Details
| Gene Symbol: MED17_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_319901.3 | XP_319901.2 | 2 - 3 | 1856 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED17_ANOGA| Gene Symbol: MED17_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_319901.3 | XP_319901.2 | 2 - 3 | 1856 | 93 | |
Set Membership: |
|