Assay Details
| Gene Symbol: ARPC2_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_319914.3 | XP_319914.2 | 5 - 6 | 781 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARPC2_ANOGA| Gene Symbol: ARPC2_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_319914.3 | XP_319914.2 | 5 - 6 | 781 | 94 | |
Set Membership: |
|