Assay Details
| Gene Symbol: 1280287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_320122.6 | XP_320122.5 | 2 - 3 | 1270 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

1280287| Gene Symbol: 1280287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_320122.6 | XP_320122.5 | 2 - 3 | 1270 | 103 | |
Set Membership: |
|