Assay Details
| Gene Symbol: INR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_320130.3 | XP_320130.3 | 11 - 12 | 3626 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

INR| Gene Symbol: INR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_320130.3 | XP_320130.3 | 11 - 12 | 3626 | 86 | |
Set Membership: |
|