Assay Details
| Gene Symbol: CPR138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_556672.3 | XP_556672.3 | 3 - 4 | 812 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CPR138| Gene Symbol: CPR138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_556672.3 | XP_556672.3 | 3 - 4 | 812 | 84 | |
Set Membership: |
|