Assay Details
| Gene Symbol: 3292554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_561090.4 | XP_561090.4 | 1 - 2 | 268 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

3292554| Gene Symbol: 3292554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_561090.4 | XP_561090.4 | 1 - 2 | 268 | 128 | |
Set Membership: |
|