Assay Details
| Gene Symbol: 3292668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_561572.4 | XP_561572.4 | 1 - 2 | 393 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

3292668| Gene Symbol: 3292668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_561572.4 | XP_561572.4 | 1 - 2 | 393 | 98 | |
Set Membership: |
|