Assay Details
| Gene Symbol: 1275382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001687731.1 | XP_001687783.1 | 1 - 2 | 398 | 95 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

1275382| Gene Symbol: 1275382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001687731.1 | XP_001687783.1 | 1 - 2 | 398 | 95 | |