Assay Details
Gene Symbol: 1268694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001687754.1 | XP_001687806.1 | 9 - 10 | 3084 | 124 | |
XM_307265.4 | XP_307265.4 | 8 - 9 | 3152 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
