Assay Details
| Gene Symbol: LANB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001687825.1 | XP_001687877.1 | 6 - 7 | 4237 | 74 | |
| XM_308240.4 | XP_308240.3 | 5 - 6 | 4417 | 74 | ||
More Information
Set Membership: |
|

LANB2
Genomic Map