Assay Details
| Gene Symbol: CPR140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001688066.1 | XP_001688118.1 | 2 - 3 | 2648 | 69 | |
| XM_565330.3 | XP_565330.3 | 3 - 4 | 2574 | 69 | ||
More Information
Set Membership: |
|

CPR140
Genomic Map