Assay Details
| Gene Symbol: CPR128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_566226.2 | XP_566226.2 | 2 - 3 | 351 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CPR128| Gene Symbol: CPR128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_566226.2 | XP_566226.2 | 2 - 3 | 351 | 85 | |
Set Membership: |
|