Assay Details
| Gene Symbol: 1275381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001688528.1 | XP_001688580.1 | 1 - 2 | 406 | 82 | |
| XM_558004.3 | XP_558004.3 | 2 - 3 | 256 | 82 | ||
More Information
Set Membership: |
|

1275381
Genomic Map