Assay Details
| Gene Symbol: AST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001689131.1 | XP_001689183.1 | 3 - 4 | 457 | 79 | |
| XM_319324.4 | XP_319324.4 | 2 - 3 | 394 | 79 | ||
More Information
Set Membership: |
|

AST2
Genomic Map