Assay Details
| Gene Symbol: HPX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003435692.1 | XP_003435740.1 | 1 - 2 | 156 | 91 | |
| XM_308561.4 | XP_308561.4 | 7 - 8 | 1934 | 91 | ||
More Information
Set Membership: |
|

HPX4
Genomic Map