Assay Details
| Gene Symbol: HPX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003435692.1 | XP_003435740.1 | 2 - 3 | 326 | 100 | |
| XM_308561.4 | XP_308561.4 | 8 - 9 | 2104 | 100 | ||
More Information
Set Membership: |
|

HPX4
Genomic Map