Assay Details
| Gene Symbol: OBP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003435718.1 | XP_003435766.1 | 2 - 3 | 236 | 74 | |
| XM_321972.3 | XP_321972.3 | 2 - 3 | 239 | 74 | ||
More Information
Set Membership: |
|

OBP10
Genomic Map