Assay Details
| Gene Symbol: OBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003435771.1 | XP_003435819.1 | 2 - 3 | 407 | 103 | |
| XM_321724.3 | XP_321724.3 | 2 - 3 | 226 | 103 | ||
More Information
Set Membership: |
|

OBP3
Genomic Map