Assay Details
| Gene Symbol: SHAKB_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003435802.1 | XP_003435850.1 | 3 - 4 | 489 | 90 | |
| XM_321635.2 | XP_321635.1 | 2 - 3 | 525 | 90 | ||
More Information
Set Membership: |
|

SHAKB_ANOGA
Genomic Map