Assay Details
| Gene Symbol: CPR126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001237304.3 | XP_001237305.3 | 3 - 4 | 773 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CPR126| Gene Symbol: CPR126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001237304.3 | XP_001237305.3 | 3 - 4 | 773 | 85 | |
Set Membership: |
|