Assay Details
| Gene Symbol: CPR116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003436418.1 | XP_003436466.1 | 1 - 2 | 283 | 57 | |
| XM_311664.5 | XP_311664.4 | 2 - 3 | 278 | 57 | ||
More Information
Set Membership: |
|

CPR116
Genomic Map