Assay Details
| Gene Symbol: CPR127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003436927.1 | XP_003436975.1 | 3 - 4 | 657 | 74 | |
| XM_310772.5 | XP_310772.4 | 3 - 4 | 573 | 74 | ||
More Information
Set Membership: |
|

CPR127
Genomic Map