Assay Details
| Gene Symbol: 1268691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_307259.4 | XP_307259.4 | 2 - 3 | 1324 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

1268691| Gene Symbol: 1268691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_307259.4 | XP_307259.4 | 2 - 3 | 1324 | 118 | |
Set Membership: |
|