Assay Details
| Gene Symbol: 1268693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_307262.4 | XP_307262.4 | 1 - 2 | 1703 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

1268693| Gene Symbol: 1268693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_307262.4 | XP_307262.4 | 1 - 2 | 1703 | 63 | |
Set Membership: |
|