Assay Details
| Gene Symbol: OBP47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_308532.3 | XP_308532.3 | 1 - 2 | 209 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OBP47| Gene Symbol: OBP47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_308532.3 | XP_308532.3 | 1 - 2 | 209 | 74 | |
Set Membership: |
|