Assay Details
| Gene Symbol: CPR55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_308909.2 | XP_308909.2 | 1 - 2 | 250 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CPR55| Gene Symbol: CPR55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_308909.2 | XP_308909.2 | 1 - 2 | 250 | 72 | |
Set Membership: |
|