Assay Details
| Gene Symbol: CPR58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_308913.4 | XP_308913.3 | 2 - 3 | 306 | 86 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CPR58| Gene Symbol: CPR58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_308913.4 | XP_308913.3 | 2 - 3 | 306 | 86 | |