Assay Details
| Gene Symbol: ARM_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_309245.5 | XP_309245.5 | 5 - 6 | 2304 | 77 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARM_ANOGA| Gene Symbol: ARM_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_309245.5 | XP_309245.5 | 5 - 6 | 2304 | 77 | |