Assay Details
| Gene Symbol: 1270953 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_309689.4 | XP_309689.4 | 2 - 3 | 265 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

1270953| Gene Symbol: 1270953 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_309689.4 | XP_309689.4 | 2 - 3 | 265 | 114 | |
Set Membership: |
|