Assay Details
| Gene Symbol: 1271336 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_310115.4 | XP_310115.3 | 2 - 3 | 698 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

1271336| Gene Symbol: 1271336 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_310115.4 | XP_310115.3 | 2 - 3 | 698 | 98 | |
Set Membership: |
|