Assay Details
| Gene Symbol: 1271338 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_310117.4 | XP_310117.3 | 1 - 2 | 43 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

1271338| Gene Symbol: 1271338 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_310117.4 | XP_310117.3 | 1 - 2 | 43 | 74 | |
Set Membership: |
|