Assay Details
| Gene Symbol: 1271342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_310122.4 | XP_310122.4 | 1 - 2 | 2358 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

1271342| Gene Symbol: 1271342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_310122.4 | XP_310122.4 | 1 - 2 | 2358 | 59 | |
Set Membership: |
|