Assay Details
| Gene Symbol: CPR137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001237741.2 | XP_001237742.2 | 1 - 2 | 148 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CPR137| Gene Symbol: CPR137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001237741.2 | XP_001237742.2 | 1 - 2 | 148 | 70 | |
Set Membership: |
|